siRNA Details
| Gene Symbol: Tspan11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024262.1 | NP_001019433.1 | 3, 4 | 400 | ||
| XM_008763291.1 | XP_008761513.1 | 3, 4 | 538 | |||
| GenBank mRNA | BC092652.1 | - | 3, 4 | 400 | 7320221 | |

Tspan11
Genomic Map