siRNA Details
| Gene Symbol: Sox5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271267.1 | NP_001258196.1 | 10, 11 | 1409 | ||
| XM_006237604.2 | XP_006237666.1 | 10, 11 | 1458 | |||
| XM_008763349.1 | XP_008761571.1 | 10, 11 | 1429 | |||
| XM_008763350.1 | XP_008761572.1 | 3, 4 | 340 | |||
| GenBank mRNA | BC086415.1 | - | 3, 4 | 343 | 7131922 | |

Sox5
Genomic Map