siRNA Details
| Gene Symbol: Cyld-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763560.1 | XP_008761782.1 | 1 | 2019 | ||
| Gene Symbol: Cyld | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017380.1 | NP_001017380.1 | 9, 10 | 2006 | ||
| XM_008772408.1 | XP_008770630.1 | 9, 10 | 3255 | |||
| GenBank mRNA | BC082001.1 | - | 9, 10 | 2006 | ||

Cyld-ps1
Genomic Map