siRNA Details
| Gene Symbol: Tmem222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113780.1 | NP_001107252.1 | 4 | 395 | ||
| GenBank mRNA | BC158875.1 | - | 4 | 395 | 7934037 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem222| Gene Symbol: Tmem222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113780.1 | NP_001107252.1 | 4 | 395 | ||
| GenBank mRNA | BC158875.1 | - | 4 | 395 | 7934037 | |