siRNA Details
| Gene Symbol: Tbc1d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107933.1 | NP_001101403.1 | 7 | 1675 | ||
| XM_006238056.2 | XP_006238118.1 | 9 | 3278 | |||
| GenBank mRNA | BC168982.1 | - | 9 | 1883 | 9030533 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbc1d2| Gene Symbol: Tbc1d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107933.1 | NP_001101403.1 | 7 | 1675 | ||
| XM_006238056.2 | XP_006238118.1 | 9 | 3278 | |||
| GenBank mRNA | BC168982.1 | - | 9 | 1883 | 9030533 | |