siRNA Details
| Gene Symbol: Exosc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107936.1 | NP_001101406.1 | 3 | 639 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Exosc3| Gene Symbol: Exosc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107936.1 | NP_001101406.1 | 3 | 639 | ||