siRNA Details
| Gene Symbol: Plekhm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191767.1 | NP_001178696.1 | 19 | 2879 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plekhm2| Gene Symbol: Plekhm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191767.1 | NP_001178696.1 | 19 | 2879 | ||