siRNA Details
| Gene Symbol: Nphp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037650.2 | NP_001032739.2 | 8 | 1319 | ||
| XM_006239464.2 | XP_006239526.1 | 8 | 8991 | |||
| XM_006239465.2 | XP_006239527.1 | 7 | 1031 | |||
| XM_006239467.2 | XP_006239529.1 | 8 | 2376 | |||
| XM_008764358.1 | XP_008762580.1 | 9 | 2506 | |||
| XM_008764359.1 | XP_008762581.1 | 8 | 8991 | |||

Nphp4
Genomic Map