siRNA Details
| Gene Symbol: RGD1311517 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014072.1 | NP_001014094.1 | 4 | 371 | ||
| XM_006239579.2 | XP_006239641.1 | 3 | 554 | |||
| XM_006239582.2 | XP_006239644.1 | 3 | 554 | |||
| XM_008764362.1 | XP_008762584.1 | 3 | 556 | |||
| XM_008764363.1 | XP_008762585.1 | 4 | 667 | |||
| XM_008764364.1 | XP_008762586.1 | 4 | 524 | |||
| XM_008764365.1 | XP_008762587.1 | 3 | 413 | |||
| XM_008764366.1 | XP_008762588.1 | 3 | 160 | |||
| GenBank mRNA | BC083613.1 | - | 4 | 371 | 7133773 | |

RGD1311517
Genomic Map