siRNA Details
| Gene Symbol: Galnt14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012109.1 | NP_001012109.1 | 6 | 830 | ||
| XM_006239775.2 | XP_006239837.1 | 6 | 997 | |||
| GenBank mRNA | BC079128.1 | - | 6 | 830 | 7127183 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Galnt14| Gene Symbol: Galnt14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012109.1 | NP_001012109.1 | 6 | 830 | ||
| XM_006239775.2 | XP_006239837.1 | 6 | 997 | |||
| GenBank mRNA | BC079128.1 | - | 6 | 830 | 7127183 | |