siRNA Details
| Gene Symbol: E2f6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100717.1 | NP_001094187.1 | 3, 4 | 383 | ||
| XM_006239913.2 | XP_006239975.1 | 4, 5 | 430 | |||
| GenBank mRNA | AY211263.1 | - | 3, 4 | 303 | ||
| AY211264.1 | - | 4, 5 | 369 | |||
| FQ216870.1 | - | 3, 4 | 629 | |||

E2f6
Genomic Map