siRNA Details
Gene Symbol: Klhl15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108021.1 | NP_001101491.1 | 2 | 418 | ||
XM_006257022.2 | XP_006257084.1 | 5 | 693 | |||
XM_006257023.2 | XP_006257085.1 | 2 | 818 | |||
XM_006257024.2 | XP_006257086.1 | 3 | 869 | |||
XM_006257027.2 | XP_006257089.1 | 2 | 1747 | |||
XM_008773257.1 | XP_008771479.1 | 1 | 2538 | |||
XM_008773259.1 | XP_008771481.1 | 4 | 913 | |||
XM_008773260.1 | XP_008771482.1 | 1 | 2538 |

More Information
Other Species Targeted

