siRNA Details
| Gene Symbol: Wdr89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014078.1 | NP_001014100.1 | 2 | 208 | ||
| XM_006240232.2 | XP_006240294.1 | 3 | 507 | |||
| XM_006240233.2 | XP_006240295.1 | 3 | 247 | |||
| XM_006240235.2 | XP_006240297.2 | 2 | 204 | |||
| GenBank mRNA | BC089849.1 | - | 2 | 208 | 7375058 | |

Wdr89
Genomic Map