siRNA Details
| Gene Symbol: Zbtb25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199496.1 | NP_955790.1 | 3 | 1108 | ||
| XM_006240236.2 | XP_006240298.1 | 2 | 1074 | |||
| XM_006240237.2 | XP_006240299.1 | 3 | 1167 | |||
| XM_008764768.1 | XP_008762990.1 | 2 | 1154 | |||
| GenBank mRNA | BC061829.1 | - | 3 | 1108 | 5599733 | |

Zbtb25
Genomic Map