siRNA Details
Gene Symbol: Zbtb25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199496.1 | NP_955790.1 | 3 | 677 | ||
XM_006240236.2 | XP_006240298.1 | 2 | 643 | |||
XM_006240237.2 | XP_006240299.1 | 3 | 736 | |||
XM_008764768.1 | XP_008762990.1 | 2 | 723 | |||
GenBank mRNA | BC061829.1 | - | 3 | 677 | 5599733 |
