siRNA Details
Gene Symbol: Zc2hc1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014079.1 | NP_001014101.1 | 2 | 915 | ||
GenBank mRNA | BC078967.1 | - | 2 | 915 | 7111557 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zc2hc1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014079.1 | NP_001014101.1 | 2 | 915 | ||
GenBank mRNA | BC078967.1 | - | 2 | 915 | 7111557 |