siRNA Details
| Gene Symbol: Serpina1f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108054.1 | NP_001101524.1 | 3 | 584 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Serpina1f| Gene Symbol: Serpina1f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108054.1 | NP_001101524.1 | 3 | 584 | ||