siRNA Details
| Gene Symbol: Wdr20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100894.1 | NP_001094364.1 | 3 | 1242 | ||
| XM_006240583.2 | XP_006240645.1 | 3 | 1178 | |||
| XM_006240585.2 | XP_006240647.1 | 2 | 995 | |||
| XM_006240586.2 | XP_006240648.1 | 2 | 995 | |||
| XM_008764933.1 | XP_008763155.1 | 4 | 1372 | |||
| XM_008764934.1 | XP_008763156.1 | 4 | 1321 | |||
| GenBank mRNA | BC091299.1 | - | 1 | 64 | ||

Wdr20
Genomic Map