siRNA Details
| Gene Symbol: Wiz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108064.1 | NP_001101534.1 | 6 | 1730 | ||
| XM_006241061.1 | XP_006241123.1 | 8 | 4782 | |||
| XM_006241062.2 | XP_006241124.1 | 8 | 4693 | |||
| XM_008765161.1 | XP_008763383.1 | 10 | 4932 | |||
| XM_008765162.1 | XP_008763384.1 | 10 | 4909 | |||
| XM_008765163.1 | XP_008763385.1 | 10 | 4467 | |||
| XM_008765164.1 | XP_008763386.1 | 10 | 4459 | |||
| XM_008765166.1 | XP_008763388.1 | 8 | 4738 | |||
| XM_008765167.1 | XP_008763389.1 | 8 | 2754 | |||
| XM_008765168.1 | XP_008763390.1 | 6 | 2193 | |||

Wiz
Genomic Map