siRNA Details
| Gene Symbol: Prtn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024264.1 | NP_001019435.1 | 5 | 687 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prtn3| Gene Symbol: Prtn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024264.1 | NP_001019435.1 | 5 | 687 | ||