siRNA Details
| Gene Symbol: Slc39a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008356.1 | NP_001008357.1 | 3 | 379 | ||
| GenBank mRNA | BC086411.1 | - | 3 | 379 | 7111777 | |
| FQ221542.1 | - | 3 | 424 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc39a3| Gene Symbol: Slc39a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008356.1 | NP_001008357.1 | 3 | 379 | ||
| GenBank mRNA | BC086411.1 | - | 3 | 379 | 7111777 | |
| FQ221542.1 | - | 3 | 424 | |||