siRNA Details
| Gene Symbol: Spic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108080.1 | NP_001101550.1 | 5 | 603 | ||
| XM_006241223.2 | XP_006241285.1 | 6 | 782 | |||
| XM_006241224.2 | XP_006241286.1 | 6 | 759 | |||
| XM_006241225.2 | XP_006241287.1 | 5 | 686 | |||
| XM_006241226.2 | XP_006241288.1 | 5 | 662 | |||
| GenBank mRNA | FQ233971.1 | - | 6 | 713 | ||

Spic
Genomic Map