siRNA Details
| Gene Symbol: Actr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108081.1 | NP_001101551.1 | 5, 6 | 580 | ||
| XM_006241243.1 | XP_006241305.1 | 5, 6 | 589 | |||
| GenBank mRNA | BC166860.1 | - | 5, 6 | 556 | 9101114 | |
| FQ220136.1 | - | 2, 3 | 225 | |||

Actr6
Genomic Map