siRNA Details
| Gene Symbol: Nab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134874.1 | NP_001128346.1 | 2 | 366 | ||
| XM_006241459.2 | XP_006241521.1 | 2 | 1874 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nab2| Gene Symbol: Nab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134874.1 | NP_001128346.1 | 2 | 366 | ||
| XM_006241459.2 | XP_006241521.1 | 2 | 1874 | |||