siRNA Details
| Gene Symbol: Phf20l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271439.1 | NP_001258368.1 | 1015 | |||
| XM_008765533.1 | XP_008763755.1 | 7 | 810 | |||
| XM_008765534.1 | XP_008763756.1 | 6 | 737 | |||
| XM_008765535.1 | XP_008763757.1 | 5 | 659 | |||
| XM_008765536.1 | XP_008763758.1 | 7 | 811 | |||
| XM_008765537.1 | XP_008763759.1 | 5 | 659 | |||
| XM_008765538.1 | XP_008763760.1 | 4 | 438 | |||
| GenBank mRNA | BC096896.1 | - | 7 | 811 | ||

Phf20l1
Genomic Map