siRNA Details
Gene Symbol: RGD1310852 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025007.1 | NP_001020178.1 | 6 | 1155 | ||
GenBank mRNA | BC091232.1 | - | 6 | 1155 | 7322456 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1310852 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025007.1 | NP_001020178.1 | 6 | 1155 | ||
GenBank mRNA | BC091232.1 | - | 6 | 1155 | 7322456 |