siRNA Details
| Gene Symbol: Fbxo43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012117.1 | NP_001012117.2 | 2 | 962 | ||
| XM_006241535.2 | XP_006241597.1 | 2 | 720 | |||
| GenBank mRNA | BC082107.1 | - | 2 | 962 | 7134870 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxo43| Gene Symbol: Fbxo43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012117.1 | NP_001012117.2 | 2 | 962 | ||
| XM_006241535.2 | XP_006241597.1 | 2 | 720 | |||
| GenBank mRNA | BC082107.1 | - | 2 | 962 | 7134870 | |