siRNA Details
| Gene Symbol: LOC680989 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008765465.1 | XP_008763687.1 | 2 | 968 | ||
| Gene Symbol: Atmin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191786.1 | NP_001178715.1 | 4 | 888 | ||
| GenBank mRNA | BC105857.1 | - | 1 | 102 | ||

LOC680989
Genomic Map