siRNA Details
| Gene Symbol: Slc45a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006241741.2 | XP_006241803.1 | 6 | 2260 | ||
| XM_008765604.1 | XP_008763826.1 | 5 | 2544 | |||
| XM_008765605.1 | XP_008763827.1 | 6 | 4515 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc45a4| Gene Symbol: Slc45a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006241741.2 | XP_006241803.1 | 6 | 2260 | ||
| XM_008765604.1 | XP_008763826.1 | 5 | 2544 | |||
| XM_008765605.1 | XP_008763827.1 | 6 | 4515 | |||