siRNA Details
| Gene Symbol: Gsdmd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130553.1 | NP_001124025.1 | 10 | 1259 | ||
| XM_006241809.2 | XP_006241871.1 | 10 | 1914 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gsdmd| Gene Symbol: Gsdmd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130553.1 | NP_001124025.1 | 10 | 1259 | ||
| XM_006241809.2 | XP_006241871.1 | 10 | 1914 | |||