siRNA Details
| Gene Symbol: Atf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100895.1 | NP_001094365.1 | 5 | 615 | ||
| GenBank mRNA | BC091222.1 | - | 5 | 589 | ||
| BC105853.1 | - | 5 | 612 | |||
| BC168224.1 | - | 6 | 667 | |||
| CO565801.1 | - | 666 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atf1| Gene Symbol: Atf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100895.1 | NP_001094365.1 | 5 | 615 | ||
| GenBank mRNA | BC091222.1 | - | 5 | 589 | ||
| BC105853.1 | - | 5 | 612 | |||
| BC168224.1 | - | 6 | 667 | |||
| CO565801.1 | - | 666 | ||||