siRNA Details
| Gene Symbol: Hoxc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750414.3 | XP_003750462.1 | 1 | 801 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hoxc10| Gene Symbol: Hoxc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750414.3 | XP_003750462.1 | 1 | 801 | ||