siRNA Details
| Gene Symbol: Piwil4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271133.1 | NP_001258062.1 | 17 | 2172 | ||
| XM_008765980.1 | XP_008764202.1 | 18 | 2383 | |||
| GenBank mRNA | BC098798.1 | - | 9 | 1045 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Piwil4| Gene Symbol: Piwil4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271133.1 | NP_001258062.1 | 17 | 2172 | ||
| XM_008765980.1 | XP_008764202.1 | 18 | 2383 | |||
| GenBank mRNA | BC098798.1 | - | 9 | 1045 | ||