siRNA Details
| Gene Symbol: Vps11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108138.1 | NP_001101608.1 | 13 | 2279 | ||
| XM_006242937.2 | XP_006242999.1 | 13 | 2261 | |||
| GenBank mRNA | BC168871.1 | - | 13 | 2114 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vps11| Gene Symbol: Vps11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108138.1 | NP_001101608.1 | 13 | 2279 | ||
| XM_006242937.2 | XP_006242999.1 | 13 | 2261 | |||
| GenBank mRNA | BC168871.1 | - | 13 | 2114 | ||