siRNA Details
| Gene Symbol: Foxr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006242968.2 | XP_006243030.1 | 1, 2 | 476 | ||
| XM_006242969.2 | XP_006243031.1 | 1, 2 | 476 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Foxr1| Gene Symbol: Foxr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006242968.2 | XP_006243030.1 | 1, 2 | 476 | ||
| XM_006242969.2 | XP_006243031.1 | 1, 2 | 476 | |||