siRNA Details
Gene Symbol: Dscaml1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108141.1 | NP_001101611.1 | 3 | 681 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
