siRNA Details
| Gene Symbol: Imp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108152.1 | NP_001101622.1 | 1 | 345 | ||
| GenBank mRNA | BC166985.1 | - | 351 | 7322293 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Imp3| Gene Symbol: Imp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108152.1 | NP_001101622.1 | 1 | 345 | ||
| GenBank mRNA | BC166985.1 | - | 351 | 7322293 | ||