siRNA Details
Gene Symbol: Cd276 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182824.2 | NP_877976.1 | 3 | 338 | ||
GenBank mRNA | AY190319.1 | - | 3 | 341 | ||
BC100064.1 | - | 3 | 338 | 7104832 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cd276 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182824.2 | NP_877976.1 | 3 | 338 | ||
GenBank mRNA | AY190319.1 | - | 3 | 341 | ||
BC100064.1 | - | 3 | 338 | 7104832 |