siRNA Details
| Gene Symbol: Cln6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191794.1 | NP_001178723.1 | 2, 3 | 199 | ||
| XM_006243238.2 | XP_006243300.1 | 3, 4 | 289 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cln6| Gene Symbol: Cln6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191794.1 | NP_001178723.1 | 2, 3 | 199 | ||
| XM_006243238.2 | XP_006243300.1 | 3, 4 | 289 | |||