siRNA Details
| Gene Symbol: Tbc1d2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108175.1 | NP_001101645.1 | 6 | 792 | ||
| XM_006243528.2 | XP_006243590.1 | 5 | 971 | |||
| XM_008766449.1 | XP_008764671.1 | 5 | 994 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbc1d2b| Gene Symbol: Tbc1d2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108175.1 | NP_001101645.1 | 6 | 792 | ||
| XM_006243528.2 | XP_006243590.1 | 5 | 971 | |||
| XM_008766449.1 | XP_008764671.1 | 5 | 994 | |||