siRNA Details
| Gene Symbol: Cnot10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007003.1 | NP_001007004.1 | 12 | 1818 | ||
| GenBank mRNA | BC083782.1 | - | 12 | 1818 | 7096219 | |
| FQ218254.1 | - | 10 | 1563 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cnot10| Gene Symbol: Cnot10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007003.1 | NP_001007004.1 | 12 | 1818 | ||
| GenBank mRNA | BC083782.1 | - | 12 | 1818 | 7096219 | |
| FQ218254.1 | - | 10 | 1563 | |||