siRNA Details
| Gene Symbol: Med20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013178.2 | NP_001013196.1 | 2 | 175 | ||
| NM_001277367.1 | NP_001264296.1 | 2 | 175 | |||
| GenBank mRNA | BC083734.1 | - | 2 | 157 | 7191561 | |
| FN804985.1 | - | 2 | 137 | |||

Med20
Genomic Map