siRNA Details
| Gene Symbol: Frs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017382.1 | NP_001017382.1 | 5 | 619 | ||
| XM_006244436.2 | XP_006244498.1 | 6 | 1136 | |||
| XM_006244437.2 | XP_006244499.1 | 6 | 990 | |||
| GenBank mRNA | AY972083.1 | - | 5 | 619 | ||

Frs3
Genomic Map