siRNA Details
| Gene Symbol: Crispld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134963.1 | NP_001128435.1 | 5 | 1374 | ||
| XM_008763477.1 | XP_008761699.1 | 5 | 1048 | |||
| XM_008763478.1 | XP_008761700.1 | 4 | 727 | |||
| GenBank mRNA | BC169118.1 | - | 5 | 1013 | 9029822 | |

Crispld1
Genomic Map