siRNA Details
| Gene Symbol: Sp140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012133.1 | NP_001012133.1 | 3 | 738 | ||
| GenBank mRNA | BC081971.1 | - | 3 | 738 | 7125855 | |
| Gene Symbol: LOC100911733 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006245337.2 | XP_006245399.1 | 2 | 126 | ||

Sp140
Genomic Map