siRNA Details
Gene Symbol: Atg4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025711.1 | NP_001020882.1 | 1, 2 | 133 | ||
XM_006245533.2 | XP_006245595.1 | 7, 8 | 673 | |||
XM_006245534.2 | XP_006245596.1 | 6, 7 | 576 | |||
GenBank mRNA | BC098833.1 | - | 1, 2 | 133 | 7378703 |
