siRNA Details
| Gene Symbol: LOC317165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047892.1 | NP_001041357.1 | 668 | |||
| GenBank mRNA | AY539903.1 | - | 668 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC317165| Gene Symbol: LOC317165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047892.1 | NP_001041357.1 | 668 | |||
| GenBank mRNA | AY539903.1 | - | 668 | |||