siRNA Details
| Gene Symbol: Nono | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012356.1 | NP_001012356.1 | 5 | 563 | ||
| GenBank mRNA | BC089880.1 | - | 5 | 563 | 7366190 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nono| Gene Symbol: Nono | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012356.1 | NP_001012356.1 | 5 | 563 | ||
| GenBank mRNA | BC089880.1 | - | 5 | 563 | 7366190 | |