siRNA Details
| Gene Symbol: Usp51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108252.1 | NP_001101722.1 | 2 | 1040 | ||
| XM_006256768.2 | XP_006256830.1 | 4 | 1372 | |||
| XM_008773076.1 | XP_008771298.1 | 3 | 2110 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usp51| Gene Symbol: Usp51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108252.1 | NP_001101722.1 | 2 | 1040 | ||
| XM_006256768.2 | XP_006256830.1 | 4 | 1372 | |||
| XM_008773076.1 | XP_008771298.1 | 3 | 2110 | |||