siRNA Details
Gene Symbol: Shroom2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047893.3 | NP_001041358.2 | 7 | 3608 | ||
XM_006256816.2 | XP_006256878.1 | 7 | 3690 | |||
XM_006256817.2 | XP_006256879.1 | 7 | 3687 | |||
XM_008773126.1 | XP_008771348.1 | 6 | 3573 | |||
XM_008773127.1 | XP_008771349.1 | 3 | 1165 | |||
XM_008773128.1 | XP_008771350.1 | 3 | 2833 | |||
XM_008773129.1 | XP_008771351.1 | 3 | 2815 | |||
XM_008773130.1 | XP_008771352.1 | 3 | 2818 | |||
XM_008773131.1 | XP_008771353.1 | 3 | 1147 | |||
XM_008773132.1 | XP_008771354.1 | 3 | 1150 | |||
GenBank mRNA | AY325213.1 | - | 5 | 3299 | ||
FQ211812.1 | - | 3 | 714 |
